Indole-4-carboxylic acid ≥98%

Supplier: Matrix Scientific
Warning

012612-5G 012612-25G
101802-978EA 56.92 USD
101802-978 101802-980
Indole-4-carboxylic acid ≥98%
Indole-4-carboxylic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 213…214 °C
MDL Number: MFCD00009739
CAS Number: 2124-55-2

Order Now

Specification Test Results

Melting point 213-214°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR