4-Bromo-2-fluoropyridine ≥97%

Supplier: Matrix Scientific
Warning

012608-1G 012608-5G 012608-25G
101802-966EA 16.92 USD
101802-966 101802-968 101802-970
4-Bromo-2-fluoropyridine ≥97%
4-Bromo-2-fluoropyridine

Hazard code(s): Irritant-Harmful

Formula: C₅H₃BrFN
MW: 175.99 g/mol
Density: 1.713 g/cm³
MDL Number: MFCD04112504
CAS Number: 128071-98-7
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR