4-Amino-2,6-dichloropyridine ≥98%

Supplier: Matrix Scientific
Warning

012550-5G 012550-25G
101802-830EA 44.62 USD
101802-830 101802-832
4-Amino-2,6-dichloropyridine ≥98%
4-Amino-2,6-dichloropyridine

Hazard code(s): Irritant

Formula: C₅H₄Cl₂N₂
Melting Pt: 174…176 °C
MDL Number: MFCD00052832
CAS Number: 2587-02-2

Order Now

Specification Test Results

Melting point 174-176°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR