2-Chloro-3-methylthiophene ≥98%

Supplier: Matrix Scientific

012540-25G 012540-100G
101802-804EA 44.62 USD
101802-804 101802-806
2-Chloro-3-methylthiophene ≥98%
2-Chloro-3-methylthiophene

Hazard code(s): Irritant

Formula: C₅H₅ClS
MW: 132.61 g/mol
Boiling Pt: 153…157 °C (1013 hPa)
Density: 1.2281 g/cm³ (25 °C)
MDL Number: MFCD00130083
CAS Number: 14345-97-2
EINECS: 238-296-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR