5-Methylthiophene-2-carboxaldehyde ≥98% for synthesis
Supplier: Matrix Scientific
Synonyms:
5-Methylthiophene-2-Carbaldehyde, 5-Methyl-2-thiophenecarboxaldehyde, 5-Methylthiophene-2-carbaldehyde
012538-25G
012538-100G
101802-800EA
49.23
USD
101802-800
101802-802
5-Methylthiophene-2-carboxaldehyde ≥98% for synthesis
5-Methylthiophene-2-carboxaldehyde
Hazard code(s): Irritant, Air Sensitive, Store in Freezer
Formula:
C₆H₆OS Boiling Pt: 92…93 °C (12mm) Density: 1.180 g/cm³ |
MDL Number:
MFCD00005434 CAS Number: 13679-70-4 EINECS: 237-178-6 |
Specification Test Results
Boiling point | 92-93°C/12m |
Density | 1.18 |
ND20 | 1.583 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...