5-Methyl-2-thenoic acid ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 5-Methylthiophene-2-carboxylic acid

012537-5G 012537-25G
101802-796EA 15.38 USD
101802-796 101802-798
5-Methyl-2-thenoic acid ≥98% for synthesis
5-Methyl-2-thenoic acid

Hazard code(s): Irritant

Formula: C₆H₆O₂S
Melting Pt: 136…138 °C
MDL Number: MFCD00005439
CAS Number: 1918-79-2
EINECS: 217-640-3

Order Now

Specification Test Results

Melting point 136-138°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR