2-Bromo-3-methylthiophene ≥98%

Supplier: Matrix Scientific

012534-5G 012534-25G
101802-786EA 15.38 USD
101802-786 101802-788
2-Bromo-3-methylthiophene ≥98%
2-Bromo-3-methylthiophene

Hazard code(s): Irritant-Harmful, Store in Freezer

Formula: C₅H₅BrS
MW: 177.06 g/mol
Boiling Pt: 173…176 °C
Density: 1.5844 g/cm³ (17.5 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00059741
CAS Number: 14282-76-9
EINECS: 238-175-2
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 173-176°C
Density 1.58
ND20 1.571

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR