2-Amino-5-fluorophenol ≥95%

Supplier: Matrix Scientific

012531-5G 012531-25G 012531-100G
101802-780EA 41.54 USD
101802-780 101802-782 101802-784
2-Amino-5-fluorophenol ≥95%
2-Amino-5-fluorophenol

Hazard code(s): Irritant, Stored under Argon

Formula: C₆H₆FNO
MW: 127.12 g/mol
Melting Pt: 135…140 °C
MDL Number: MFCD00671759
CAS Number: 53981-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR