2-Amino-4-fluorophenol ≥98%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
5-Fluoro-2-hydroxyaniline
012530-5G
012530-25G
101802-776EA
23.08
USD
101802-776
101802-778
2-Amino-4-fluorophenol ≥98%
2-Amino-4-fluorophenol
Hazard code(s): Irritant
Formula:
C₆H₆FNO MW: 127.12 g/mol |
MDL Number:
MFCD00077451 CAS Number: 399-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...