2-Bromo-6-(trifluoromethyl)pyridine ≥97%
Supplier: Matrix Scientific
012527-5G
012527-25G
101802-768EA
36.92
USD
101802-768
101802-770
2-Bromo-6-(trifluoromethyl)pyridine ≥97%
2-Bromo-6-(trifluoromethyl)pyridine
Hazard code(s): Irritant
Formula:
C₆H₃BrF₃N MW: 226 g/mol Boiling Pt: 78…79 °C (14mm) Melting Pt: 45…46 °C |
MDL Number:
MFCD00153087 CAS Number: 189278-27-1 |
Specification Test Results
Boiling point | 78-79°C/14m |
Melting point | 45-46°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...