3,4,5-Trichloropyridine ≥98%

Supplier: Matrix Scientific
Warning

012525-5G 012525-25G
101802-758EA 15.38 USD
101802-758 101802-760
3,4,5-Trichloropyridine ≥98%
3,4,5-Trichloropyridine

Hazard code(s): Irritant

Formula: C₅H₂Cl₃N
Boiling Pt: 213…215 °C
Melting Pt: 75…77 °C
MDL Number: MFCD00051685
CAS Number: 33216-52-3

Order Now

Specification Test Results

Boiling point 213-215°C
Melting point 75-77°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR