4,5-Difluoroanthranilic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Amino-4,5-difluorobenzoic acid

012523-5G 012523-25G
101802-750EA 36.92 USD
101802-750 101802-752
4,5-Difluoroanthranilic acid ≥97%
4,5-Difluoroanthranilic acid

Hazard code(s): Irritant

Formula: C₇H₅F₂NO₂
MW: 173.12 g/mol
Melting Pt: 181…183 °C
MDL Number: MFCD00077509
CAS Number: 83506-93-8

Order Now

Specification Test Results

Melting point 181-183°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR