6-Bromonicotinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Bromopyridine-3-carboxylic acid, 2-Bromo-5-pyridinecarboxylic acid, 6-Bromo-3-pyridinecarboxylic acid

012519-5G 012519-25G
101802-742EA 30.77 USD
101802-742 101802-744
6-Bromonicotinic acid ≥98%
6-Bromonicotinic acid

Hazard code(s): Irritant

Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Melting Pt: 197…200 °C
MDL Number: MFCD01927100
CAS Number: 6311-35-9

Order Now

Specification Test Results

Melting point 197-200°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR