2,4-Dichloro-5-fluoro-3-nitrobenzoic acid ≥97%

Supplier: Matrix Scientific

012515-25G 012515-100G
101802-728EA 36.92 USD
101802-728 101802-730
2,4-Dichloro-5-fluoro-3-nitrobenzoic acid ≥97%
2,4-Dichloro-5-fluoro-3-nitrobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₂Cl₂FNO₄
MW: 254 g/mol
Melting Pt: 193…197 °C
MDL Number: MFCD03427137
CAS Number: 106809-14-7

Order Now

Specification Test Results

Melting point 193-197°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR