8-Quinolinecarbaldehyde
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
Quinoline-8-carbaldehyde, 8-Formylquinoline, 8-Quinolinecarboxaldehyde, Quinoline-8-carboxaldehyde
011529-1G
011529-5G
101800-824EA
60
USD
101800-824
101800-826
8-Quinolinecarbaldehyde
8-Quinolinecarbaldehyde
Formula:
C₁₀H₇NO MW: 157.17 g/mol |
MDL Number:
MFCD00805837 CAS Number: 38707-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...