Methyl propionylacetate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 3-oxovalerate, Methyl 3-oxo-pentanoate

011295-100G 011295-500G
101800-372EA 69.23 USD
101800-372 101800-374
Methyl propionylacetate ≥97%
Methyl propionylacetate

Hazard code(s): Irritant

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 73…74 °C (5mm)
Density: 1.037 g/cm³
MDL Number: MFCD00011705
CAS Number: 30414-53-0
EINECS: 250-184-3

Order Now

Specification Test Results

Boiling point 73-74°C/5mm
Density 1.037
ND20 1.422

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR