4-Chloro-3-fluoroaniline ≥98%

Supplier: Matrix Scientific

011294-25G 011294-100G
101800-368EA 52.31 USD
101800-368 101800-370
4-Chloro-3-fluoroaniline ≥98%
4-Chloro-3-fluoroaniline

Hazard code(s): Toxic

Formula: C₆H₅ClFN
Melting Pt: 59…61 °C
MDL Number: MFCD01090987
CAS Number: 367-22-6
EINECS: 206-683-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR