2-Bromo-4-(trifluoromethyl)pyridine for synthesis

Supplier: Matrix Scientific

011289-5G 011289-25G
101800-354EA 78.46 USD
101800-354 101800-356
2-Bromo-4-(trifluoromethyl)pyridine for synthesis
2-Bromo-4-(trifluoromethyl)pyridine
Formula: C₆H₃BrF₃N
MW: 226
MDL Number: MFCD00153085
CAS Number: 175205-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR