2'-Chloro-4',5'-difluoroacetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(2-Chloro-4,5-difluorophenyl)ethanone, 1-chloro-3,4-difluor-6-ethanoylbenzene

011288-5G 011288-25G 011288-100G
101800-346EA 23.08 USD
101800-346 101800-348 101800-350
2'-Chloro-4',5'-difluoroacetophenone ≥98%
2'-Chloro-4',5'-difluoroacetophenone

Hazard code(s): Irritant

Formula: C₈H₅ClF₂O
MW: 190.58 g/mol
MDL Number: MFCD00671761
CAS Number: 121872-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR