(2-Chloro-5-fluorophenyl)acetic acid ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 2-Chloro-5-fluorophenylacetic acid

011282-1G 011282-5G
101800-324EA 56.92 USD
101800-324 101800-326
(2-Chloro-5-fluorophenyl)acetic acid ≥97%
(2-Chloro-5-fluorophenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₃ClF)CH₂COOH
MW: 188.59 g/mol
Melting Pt: 114…115 °C
MDL Number: MFCD01016155
CAS Number: 177985-33-0

Order Now

Specification Test Results

Melting point 114-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR