2-Bromo-4-fluorobenzaldehyde ≥98%

Supplier: Matrix Scientific

011279-25G 011279-100G
101800-312EA 53.85 USD
101800-312 101800-314
2-Bromo-4-fluorobenzaldehyde ≥98%
2-Bromo-4-fluorobenzaldehyde

Hazard code(s): Irritant, Light Sensitive

Formula: C₇H₄BrFO
MW: 203.01 g/mol
Melting Pt: 65…68 °C
MDL Number: MFCD00672923
CAS Number: 59142-68-6

Order Now

Specification Test Results

Melting point 65-68°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR