2',6'-Dichloro-3'-fluoroacetophenone ≥98%

Supplier: Matrix Scientific

011278-25G 011278-100G
101800-308EA 24.62 USD
101800-308 101800-310
2',6'-Dichloro-3'-fluoroacetophenone ≥98%
2',6'-Dichloro-3'-fluoroacetophenone

Hazard code(s): Irritant

Formula: C₈H₅Cl₂FO
MW: 207.03 g/mol
Boiling Pt: 255 °C
Density: 1.403 g/cm³
MDL Number: MFCD02093760
CAS Number: 290835-85-7

Order Now

Specification Test Results

Boiling point 255°C
Density 1.403
ND20 1.529

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR