2-Amino-3-nitropyridine ≥99% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 3-Nitropyridin-2-ylamine, 3-Nitropyridin-2-amine, 3-Nitro-2-pyridylamine

011276-5G 011276-25G
101800-300EA 32.31 USD
101800-300 101800-302
2-Amino-3-nitropyridine ≥99% for synthesis
2-Amino-3-nitropyridine

Hazard code(s): Irritant

Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
Melting Pt: 165…167 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006314
CAS Number: 4214-75-9
EINECS: 224-144-0

Order Now

Specification Test Results

Melting point 165-167°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR