Methyl indole-4-carboxylate ≥99%

Supplier: Matrix Scientific

011275-5G 011275-25G
101800-296EA 29.23 USD
101800-296 101800-298
Methyl indole-4-carboxylate ≥99%
Methyl indole-4-carboxylate

Hazard code(s): Irritant, Store in Freezer

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 69…71 °C
MDL Number: MFCD00191222
CAS Number: 39830-66-5

Order Now

Specification Test Results

Melting point 69-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR