2-Amino-4-methyl-3-nitropyridine ≥97%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
4-Methyl-3-nitro-2-pyridylamine, 4-Methyl-3-nitropyridin-2-amine, 2-Amino-3-nitro-4-picoline
011257-5G
011257-25G
101800-244EA
29.23
USD
101800-244
101800-246
2-Amino-4-methyl-3-nitropyridine ≥97%
2-Amino-4-methyl-3-nitropyridine
Hazard code(s): Irritant
Formula:
C₆H₇N₃O₂ MW: 153.14 g/mol Melting Pt: 140 °C |
MDL Number:
MFCD00006315 CAS Number: 6635-86-5 |
Specification Test Results
Melting point | 139-141°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...