2-Chloro-3-nitropyridine ≥98%

Supplier: Matrix Scientific
Warning

011254-25G 011254-100G
101800-236EA 29.23 USD
101800-236 101800-238
2-Chloro-3-nitropyridine ≥98%
2-Chloro-3-nitropyridine

Hazard code(s): Irritant, Store in Freezer

Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 99 - 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00006232
CAS Number: 5470-18-8
EINECS: 226-799-8

Order Now

Specification Test Results

Melting point 101-103°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR