6-Aminonicotinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Aminopyridine-3-carboxylic acid, 6-Amino-3-pyridinecarboxylic acid

011253-5G 011253-25G 011253-100G
101800-230EA 15.38 USD
101800-230 101800-232 101800-234
6-Aminonicotinic acid ≥98%
6-Aminonicotinic acid

Hazard code(s): Irritant

Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: ? 300 °C
MDL Number: MFCD00006326
CAS Number: 3167-49-5
EINECS: 221-630-4
Merck Index: 13,00455

Order Now

Specification Test Results

Melting point 308-310°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR