5-Bromo-2-methylpyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-picoline, 3-Bromo-6-picoline

011252-5G 011252-25G 011252-100G
101800-224EA 15.38 USD
101800-224 101800-226 101800-228
5-Bromo-2-methylpyridine ≥98%
5-Bromo-2-methylpyridine

Hazard code(s): Irritant

Formula: C₆H₆BrN
MW: 172.02 g/mol
Melting Pt: 32…36 °C
MDL Number: MFCD00661170
CAS Number: 3430-13-5

Order Now

Specification Test Results

Melting point 84-86°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR