5-Bromo-2-iodopyridine ≥98% for synthesis

Supplier: Matrix Scientific
Warning

011251-25G 011251-100G
101800-220EA 76.92 USD
101800-220 101800-222
5-Bromo-2-iodopyridine ≥98% for synthesis
5-Bromo-2-iodopyridine

Hazard code(s): Irritant, Light Sensitive

Formula: C₅H₃BrIN
Melting Pt: 113…114 °C
MDL Number: MFCD00691506
CAS Number: 223463-13-6

Order Now

Specification Test Results

Melting point 113-114°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR