2-Bromo-5-fluoropyridine ≥98%

Supplier: Matrix Scientific

011250-25G 011250-100G
101800-218EA 67.69 USD
101800-218 102832-836
2-Bromo-5-fluoropyridine ≥98%
2-Bromo-5-fluoropyridine

Hazard code(s): Irritant

Formula: C₅H₃BrFN
MW: 175.99 g/mol
Boiling Pt: 80…83 °C (44mm)
Melting Pt: 27…29 °C
MDL Number: MFCD00234011
CAS Number: 41404-58-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 80-83°C/44m
Melting point 27-29°C
ND20 1.5396

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR