3-Bromo-4-methylpyridine ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 3-Bromo-4-picoline

011246-5G 011246-25G 011246-100G
101800-206EA 41.54 USD
101800-206 101800-208 101800-210
3-Bromo-4-methylpyridine ≥97%
3-Bromo-4-methylpyridine

Hazard code(s): Irritant

Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 199…200 °C
Density: 1.549 g/cm³ (20 °C)
MDL Number: MFCD00082592
CAS Number: 3430-22-6

Order Now

Specification Test Results

Boiling point 199-200°C
Density 1.549
ND20 1.56

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR