4-Bromo-2-chloropyridine ≥97%

Supplier: Matrix Scientific

011244-5G 011244-25G
101800-202EA 30.77 USD
101800-202 101800-204
4-Bromo-2-chloropyridine ≥97%
4-Bromo-2-chloropyridine

Hazard code(s): Irritant

Formula: C₅H₃BrClN
MW: 192.44 g/mol
Boiling Pt: 70 °C (3mm)
Melting Pt: 18…20 °C
Density: 1.734 g/cm³
MDL Number: MFCD03840756
CAS Number: 73583-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR