5-Amino-2-chloropyridine ≥98%

Supplier: Matrix Scientific
Warning

011219-25G 011219-100G
101800-152EA 21.54 USD
101800-152 101800-154
5-Amino-2-chloropyridine ≥98%
5-Amino-2-chloropyridine

Hazard code(s): Irritant

Formula: C₅H₅ClN₂
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: 83 °C
MDL Number: MFCD00006243
CAS Number: 5350-93-6
EINECS: 226-322-3

Order Now

Specification Test Results

Melting point 80-82°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR