6-Nitroindole ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Nitro-1H-indole

011217-1G 011217-5G 011217-25G
101800-142EA 15.38 USD
101800-142 101800-144 101800-146
6-Nitroindole ≥98%
6-Nitroindole

Hazard code(s): Irritant, Irritant-Harmful, Store in Freezer

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 143…144 °C
MDL Number: MFCD00051497
CAS Number: 4769-96-4
EINECS: 225-311-0

Order Now

Specification Test Results

Melting point 143-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR