(4-Iodophenyl)acetic acid ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 2-(4-Iodophenyl)acetic acid

011210-5G 011210-25G
101800-134EA 29.23 USD
101800-134 101800-136
(4-Iodophenyl)acetic acid ≥98%
(4-Iodophenyl)acetic acid

Hazard code(s): Irritant, Light Sensitive

Formula: (C₆H₄I)CH₂COOH
MW: 262.05 g/mol
Melting Pt: 134…136 °C
MDL Number: MFCD00082985
CAS Number: 1798-06-7

Order Now

Specification Test Results

Melting point 134-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR