3,5-Dihydroxybenzaldehyde ≥98%
Supplier: Matrix Scientific
011209-1G
011209-5G
101800-130EA
67.69
USD
101800-130
101800-132
3,5-Dihydroxybenzaldehyde ≥98%
3,5-Dihydroxybenzaldehyde
Hazard code(s): Irritant, Air Sensitive
Formula:
C₇H₆O₃ MW: 138.12 g/mol Melting Pt: 159…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016611 CAS Number: 26153-38-8 EINECS: 247-479-4 |
Specification Test Results
Melting point | 159-160°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...