4-Amino-3,5-dichloropyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 3,5-Dichloro-4-pyridylamine, 3,5-Dichloropyridin-4-amine

011181-25G 011181-100G
101800-104EA 30.77 USD
101800-104 101800-106
4-Amino-3,5-dichloropyridine ≥98%
4-Amino-3,5-dichloropyridine

Hazard code(s): Irritant

Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Melting Pt: 159…161 °C
MDL Number: MFCD00125023
CAS Number: 22889-78-7
EINECS: 245-304-6

Order Now

Specification Test Results

Melting point 159-161°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR