4-Amino-2-chloropyridine ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 2-Chloro-4-aminopyridine

011180-25G 011180-100G
101800-098EA 38.46 USD
101800-098 101800-100
4-Amino-2-chloropyridine ≥98%
4-Amino-2-chloropyridine

Hazard code(s): Irritant

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 87 - 89 °C
MDL Number: MFCD00060089
CAS Number: 14432-12-3
EINECS: 238-403-0

Order Now

Specification Test Results

Melting point 90-94°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR