3,5-Dibromopyridine ≥98%

Supplier: Matrix Scientific
Warning

011179-25G 011179-100G
101800-094EA 15.38 USD
101800-094 101800-096
3,5-Dibromopyridine ≥98%
3,5-Dibromopyridine

Hazard code(s): Irritant

Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 111…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00014634
CAS Number: 625-92-3
EINECS: 210-916-4

Order Now

Specification Test Results

Melting point 111-112°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR