3,5-Dichlorothioanisole ≥97%

Supplier: Matrix Scientific

011175-5G 011175-25G
101800-076EA 33.85 USD
101800-076 101800-078
3,5-Dichlorothioanisole ≥97%
3,5-Dichlorothioanisole

Hazard code(s): Irritant, Stench

Formula: C₇H₆Cl₂S
MW: 193.1 g/mol
Boiling Pt: 262…263 °C
Density: 1.349 g/cm³
MDL Number: MFCD00870149
CAS Number: 68121-46-0

Order Now

Specification Test Results

Boiling point 262-263°C
Density 1.349
ND20 1.613

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR