3-Methoxythioanisole ≥98%

Supplier: Matrix Scientific

011173-5G 011173-25G
101800-072EA 27.69 USD
101800-072 101800-074
3-Methoxythioanisole ≥98%
3-Methoxythioanisole

Hazard code(s): Irritant, Stench

Formula: C₈H₁₀OS
MDL Number: MFCD03789162
CAS Number: 2388-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR