4-Nitrothioanisole ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Nitrophenyl methyl sulphide, 1-Nitro-4-(methylthio)benzene

011171-25G 011171-100G
101800-064EA 83.08 USD
101800-064 101800-066
4-Nitrothioanisole ≥98%
4-Nitrothioanisole

Hazard code(s): Irritant, Stench

Formula: C₇H₇NO₂S
MW: 169.2 g/mol
Boiling Pt: 135…140 °C (2 mmHg)
Melting Pt: 66…68 °C
MDL Number: MFCD00010868
CAS Number: 701-57-5

Order Now

Specification Test Results

Melting point 66-68°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR