3-(Methylthio)aniline ≥97%
Supplier: Matrix Scientific
011169-25G
011169-100G
101800-056EA
30.77
USD
101800-056
101800-058
3-(Methylthio)aniline ≥97%
3-(Methylthio)aniline
Hazard code(s): Irritant, Stench
Formula:
C₇H₉NS Boiling Pt: 163…165 °C (16mm) Density: 1/130 g/cm³ |
MDL Number:
MFCD00007793 CAS Number: 1783-81-9 EINECS: 217-232-5 UN: 2810 ADR: 6.1,III |
Specification Test Results
Boiling point | 163-165°C/1 |
Density | 1/130 |
ND20 | 1.637 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...