2,3-Dimethoxypyridine ≥97%

Supplier: Matrix Scientific
Warning

011168-5G 011168-25G
101800-052EA 64.62 USD
101800-052 101800-054
2,3-Dimethoxypyridine ≥97%
2,3-Dimethoxypyridine

Hazard code(s): Irritant

Formula: C₇H₉NO₂
MW: 139.15 g/mol
Boiling Pt: 98…100 °C (16mm)
MDL Number: MFCD03095043
CAS Number: 52605-97-7

Order Now

Specification Test Results

Boiling point 98-100°C/16

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR