1-Boc-3-piperidone ≥97%

Supplier: Matrix Scientific

011166-25G 011166-100G
101800-044EA 75.38 USD
101800-044 101800-046
1-Boc-3-piperidone ≥97%
1-Boc-3-piperidone

Hazard code(s): Store in Freezer

Formula: C₁₀H₁₇NO₃
Melting Pt: 35…40 °C
MDL Number: MFCD01631193
CAS Number: 98977-36-7

Order Now

Specification Test Results

Melting point 35-40


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR