2-Amino-3,5-dibromopyridine ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 3,5-Dibromo-2-pyridylamine, 3,5-Dibromopyridin-2-amine

011157-25G 011157-100G
101800-020EA 21.54 USD
101800-020 101800-022
2-Amino-3,5-dibromopyridine ≥98%
2-Amino-3,5-dibromopyridine

Hazard code(s): Irritant

Formula: C₅H₄Br₂N₂
MW: 251.91 g/mol
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 102…104 °C
MDL Number: MFCD00038041
CAS Number: 35486-42-1
EINECS: 252-590-6

Order Now

Specification Test Results

Melting point 102-104°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR