3-Bromo-2-chlorothiophene ≥98%

Supplier: Matrix Scientific
Warning

011156-1G 011156-5G 011156-25G
101800-014EA 15.38 USD
101800-014 101800-016 101800-018
3-Bromo-2-chlorothiophene ≥98%
3-Bromo-2-chlorothiophene

Hazard code(s): Irritant

Formula: C₄H₂BrClS
Boiling Pt: 70 °C (9mm)
Density: 1.82 g/cm³ (20 °C)
MDL Number: MFCD00043883
CAS Number: 40032-73-3
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 70°C/9mm
Density 1.808
ND20 1.596


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR