Ethyl-6-chloronicotinate ≥98%

Supplier: Matrix Scientific

Synonyms: Ethyl 6-chloronicotinate

011153-25G 011153-100G
101800-004EA 38.46 USD
101800-004 101800-006
Ethyl-6-chloronicotinate ≥98%
Ethyl-6-chloronicotinate

Hazard code(s): Irritant, Store in Freezer

Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Boiling Pt: 86…87 °C (1mm)
Melting Pt: 26…30 °C
MDL Number: MFCD00082739
CAS Number: 49608-01-7

Order Now

Specification Test Results

Boiling point 86-87°C/1mm
ND20 1.522

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR