3-Amino-2-chloropyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-3-pyridylamine, 2-Chloropyridin-3-amine

011152-100G 011152-250G
101800-000EA 64.62 USD
101800-000 101800-002
3-Amino-2-chloropyridine ≥98%
3-Amino-2-chloropyridine

Hazard code(s): Irritant

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 130…134 °C (22mm)
Melting Pt: 75…78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006238
CAS Number: 6298-19-7
EINECS: 228-572-9

Order Now

Specification Test Results

Boiling point 130-134°C/2
Melting point 75-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR