2-Bromo-4-fluoro-1-iodobenzene ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 3-Bromo-1-fluoro-4-iodobenzene, 2-Bromo-4-fluoroiodobenzene, 1-Bromo-5-fluoro-2-iodobenzene

011140-25G 011140-100G
101799-984EA 60 USD
101799-984 101799-986
2-Bromo-4-fluoro-1-iodobenzene ≥98%
2-Bromo-4-fluoro-1-iodobenzene

Hazard code(s): Irritant

Formula: C₆H₃BrFI
MW: 300.9 g/mol
MDL Number: MFCD00142868
CAS Number: 202865-73-4

Order Now

Specification Test Results

ND20 1.6292

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR